Skip to Content
Merck
All Photos(1)

Key Documents

AV48196

Sigma-Aldrich

Anti-CRYAB antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-CRYA2, Anti-CTPP2, Anti-Crystallin, α B, Anti-HSPB5

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$482.00

$482.00


Check Cart for Availability


Select a Size

Change View
100 μL
$482.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

$482.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

20 kDa

species reactivity

rat, mouse, human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CRYAB(1410)

General description

Crystallin, α B (CRYAB) is a molecular chaperone that holds protein in soluble aggregates. It functions as a tumor suppressor by interacting with adherens junction to inhibit the progression of nasopharyngeal carcinoma. It is also known to inhibit stimulation of CD4+ T cells in relapsing-remitting multiple sclerosis patients.
Rabbit Anti-CRYAB antibody recognizes canine, rabbit, pig, human, mouse, rat, and bovine CRYAB.

Immunogen

Synthetic peptide directed towards the C terminal region of human CRYAB

Application

Rabbit Anti-CRYAB antibody is suitable for western blot applications at a concentration of 5 μg/ml.

Biochem/physiol Actions

Alpha crystallins are composed of: alpha-A and alpha-B, for acidic and basic, respectively. They act as molecular chaperones although they do not renature proteins and release them in the fashion of a true chaperone; instead they hold them in large soluble aggregates. Post-translational modifications decrease the ability to chaperone. Two additional functions of alpha crystallins are an autokinase activity and participation in the intracellular architecture. Alpha-A and alpha-B are differentially expressed; alpha-A is preferentially restricted to the lens and alpha-B is expressed widely in many tissues and organs. Elevated expression of alpha-B crystallin occurs in many neurological diseases; a missense mutation cosegregated in a family with a desmin-related myopathy.

Sequence

Synthetic peptide located within the following region: KYRIPADVDPLTITSSLSSDGVLTVNGPRKQVSGPERTIPITREEKPAVT

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Que Lan Quach et al.
Multiple sclerosis (Houndmills, Basingstoke, England), 19(14), 1867-1877 (2013-06-06)
Suppression of activation of pathogenic CD4(+) T cells is a potential therapeutic intervention in multiple sclerosis (MS). We previously showed that a small heat shock protein, CRYAB, reduced T cell proliferation, pro-inflammatory cytokine production and clinical signs of experimental allergic
Z Huang et al.
Oncogene, 31(32), 3709-3720 (2011-12-14)
Alpha B-crystallin (CRYAB) maps within the nasopharyngeal carcinoma (NPC) tumor-suppressive critical region 11q22-23 and its downregulation is significantly associated with the progression of NPC. However, little is known about the functional impact of CRYAB on NPC progression. In this study

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service