Skip to Content
Merck
All Photos(3)

Key Documents

AV47897

Sigma-Aldrich

Anti-EXD antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-CG8933, Anti-DExd, Anti-Dm-EXD, Anti-Dmel_CG8933, Anti-Dpbx, Anti-EXD, Anti-Td48

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$482.00

$482.00


Check Cart for Availability


Select a Size

Change View
100 μL
$482.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$482.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

42 kDa

species reactivity

human, rat

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Drosophila melanogaster ... exd(32567)

General description

Extradenticle (EXD) is a Drosophila transcription factor that regulates brain and eye development. It modulates the transcription network in the dorsal retinal rim and the FGF/branchless expression in mesodermal bridge cells.
Rabbit Anti-EXD antibody recognizes mouse, and rat EXD.

Immunogen

Synthetic peptide corresponding to a region of Fruit fly

Application

Rabbit Anti-EXD antibody is suitable for western blot applications at a concentration of 5μg/ml.

Biochem/physiol Actions

As a transcription factor, exd acts with the selector homeodomain proteins altering the regulation of downstream target genes such as wingless, teashirt and decapentaplegic. Thus exd affects segmental identity.

Sequence

Synthetic peptide located within the following region: EAKEELARKCGITVSQVSNWFGNKRIRYKKNIGKAQEEANLYAAKKAAGA

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mathias F Wernet et al.
Development (Cambridge, England), 141(4), 918-928 (2014-02-06)
A narrow band of ommatidia in the dorsal periphery of the Drosophila retina called the dorsal rim area (DRA) act as detectors for polarized light. The transcription factor Homothorax (Hth) is expressed in DRA inner photoreceptors R7 and R8 and
Samir Merabet et al.
EMBO reports, 6(8), 762-768 (2005-07-12)
The stereotyped outgrowth of tubular branches of the Drosophila tracheal system is orchestrated by the local and highly dynamic expression profile of branchless (bnl), which encodes a secreted fibroblast growth factor (FGF)-like molecule. Despite the importance of the spatial and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service