Skip to Content
Merck
All Photos(1)

Key Documents

AV46829

Sigma-Aldrich

Anti-UGT1A4 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-HUG-BR2, Anti-UDP glucuronosyltransferase 1 family, polypeptide A4, Anti-UDPGT, Anti-UGT1D

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

57 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... UGT1A4(54657)

Immunogen

Synthetic peptide directed towards the N terminal region of human UGT1A4

Biochem/physiol Actions

UGT1A4 is an enzyme of the glucuronidation pathway that transforms small lipophilic molecules, such as steroids, hormones, and drugs, into water-soluble, excretable metabolites. This enzyme has some glucuronidase activity towards amines, steroids, and sapogenins. This gene is part of a complex locus that encodes several UDP-glucuronosyltransferases. The locus includes thirteen unique alternate first exons followed by four common exons. Four of the alternate first exons are considered pseudogenes. Each of the remaining nine 5′ exons may be spliced to the four common exons, resulting in nine proteins with different N-termini and identical C-termini. Each first exon encodes the substrate binding site, and is regulated by its own promoter.

Sequence

Synthetic peptide located within the following region: VVLTPEVNMHIKEEKFFTLTAYAVPWTQKEFDRVTLGYTQGFFETEHLLK

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ursula Ehmer et al.
Hepatology (Baltimore, Md.), 55(6), 1912-1921 (2012-01-04)
Gilbert syndrome (GS) is characterized by intermittent unconjugated hyperbilirubinemia without structural liver damage, affecting about 10% of the white population. In GS the UGT1A1*28 variant reduces bilirubin conjugation by 70% and is associated with irinotecan and protease inhibitor side effects.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service