Skip to Content
Merck
All Photos(3)

Key Documents

AV46079

Sigma-Aldrich

Anti-PPAT antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-ATASE, Anti-GPAT, Anti-PRAT, Anti-Phosphoribosyl pyrophosphate amidotransferase

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$716.00

$716.00


Check Cart for Availability


Select a Size

Change View
100 μL
$716.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$716.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

56 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PPAT(5471)

General description

Phosphoribosyl pyrophosphate amidotransferase/glutamine phosphoribosyl pyrophosphate amidotransferase (PPAT, PRAT, ATASE, GPAT) catalyzes the first committed step in de novo purine biosynthesis by converting α-phosphoribosylpyrophosphate (α-PRPP) into 5-β-phosphoribosylamine. Phosphoribosylamine (PRA) is the first intermediate in the common purine/thiamine biosynthetic pathway.

Specificity

Anti-PPAT polyclonal antibody reacts with bovine, rat, canine, and human phosphoribosyl pyrophosphate amidotransferase/glutamine phosphoribosyl pyrophosphate amidotransferase enzymes.

Immunogen

Synthetic peptide directed towards the N terminal region of human PPAT

Application

Anti-PPAT polyclonal antibody is used to tag phosphoribosyl pyrophosphate amidotransferase for detection and quantitation by Western blotting and in plasma by immunohistochemical (IHC) techniques. It is used as a probe to determine the roles of phosphoribosyl pyrophosphate amidotransferase in purine/pyrimidine biosynthesis.

Biochem/physiol Actions

PPAT is a member of the purine/pyrimidine phosphoribosyltransferase family. This protein is a regulatory allosteric enzyme that catalyzes the first step of de novo purine nucleotide biosynthesis. Its gene and PAICS/AIRC, a bifunctional enzyme catalyzing steps six and seven in the purine nucleotide biosynthesis pathway, are located in close proximity on chromosome 4.The protein encoded by this gene is a member of the purine/pyrimidine phosphoribosyltransferase family. This protein is a regulatory allosteric enzyme that catalyzes the first step of de novo purine nucleotide biosynthesis. This gene and PAICS/AIRC, a bifunctional enzyme catalyzing steps six and seven in the purine nucleotide biosynthesis pathway, are located in close proximity on chromosome 4.

Sequence

Synthetic peptide located within the following region: VTSDGSSVPTFKSHKGMGLVNHVFTEDNLKKLYVSNLGIGHTRYATTGKC

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service