Skip to Content
Merck
All Photos(2)

Documents

AV45597

Sigma-Aldrich

Anti-NR0B1 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-AHC, Anti-AHCH, Anti-AHX, Anti-DAX-1, Anti-DAX1, Anti-DSS, Anti-GTD, Anti-Nuclear receptor subfamily 0, group B, member 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

52 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... NR0B1(190)

Immunogen

Synthetic peptide directed towards the N terminal region of human NR0B1

Application

Anti-NR0B1 antibody produced in rabbit is suitable for western blotting at a concentration of 2.5μg/ml and for immunohistochemistry of paraffin-embedded tissue sections at a concentration of 4-8μg/ml.

Biochem/physiol Actions

Nuclear receptor subfamily 0, group B, member 1 (NR0B1; DAX-1) is a DNA-binding protein that negatively regulates retinoic acid receptor-mediated transcription. It is an antagonist to the protein Sry and functions as an anti-testis protein. Mutations in gene encoding NR0B1 result in primary adrenal insufficiency or congenital adrenal hypoplasia in early childhood and hypogonadotropic hypogonadism.

Sequence

Synthetic peptide located within the following region: TSSKQTHVAPAAPEARPGGAWWDRSYFAQRPGGKEALPGGRATALLYRCC

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Claudilene Battistin et al.
Arquivos brasileiros de endocrinologia e metabologia, 56(8), 496-500 (2013-01-09)
We report a case of adrenal hypoplasia congenita (AHC) and hypogonadotropic hypogonadism (HH) due to a novel DAX1 mutation. A 19-month-old boy with hyperpigmentation and failure to thrive came to our service for investigation. Three brothers of the patient had
X Yuan et al.
The Journal of biological chemistry, 276(49), 46647-46654 (2001-10-05)
This study investigated interactions between SRY, the Y chromosome encoded male sex determining factor, and the androgen receptor (AR). Coexpression of AR and SRY caused marked repression of AR transcriptional activity on a series of androgen-responsive reporter genes. Mammalian one-
Bryan W Teets et al.
Journal of cellular physiology, 227(4), 1501-1511 (2011-06-17)
Retinoic acid (RA) is critical for embryonic development and cellular differentiation. Previous work in our laboratory has shown that blocking the RA-dependent increase in pre-β cell leukemia transcription factors (PBX) mRNA and protein levels in P19 cells prevents endodermal and
Naoki Mine et al.
Molecular cancer therapeutics, 13(9), 2215-2225 (2014-07-24)
CBP501 is an anticancer drug candidate that was investigated in two randomized phase II clinical trials for patients with nonsquamous non-small cell lung cancer (NSCLC) and malignant pleural mesothelioma (MPM). CBP501 has been shown to have two mechanisms of action

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service