Skip to Content
Merck
All Photos(1)

Key Documents

AV45038

Sigma-Aldrich

Anti-AADAC antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Arylacetamide deacetylase (esterase), Anti-DAC

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$482.00

$482.00


Check Cart for Availability


Select a Size

Change View
100 μL
$482.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$482.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

44 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... AADAC(13)

Immunogen

Synthetic peptide directed towards the C terminal region of human AADAC

Application

Anti-AADAC antibody produced in rabbit is suitable for western blotting at a concentration of 2.5μg/ml.

Biochem/physiol Actions

Arylacetamide deacetylase (AADAC) is an esterase involved in the hydrolysis of a variety of drugs such as flutamide, phenacetin and rifamycin.

Sequence

Synthetic peptide located within the following region: NYGSSELAKKYPGFLDVRAAPLLADDNKLRGLPLTYVITCQYDLLRDDGL

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Akinobu Watanabe et al.
Drug metabolism and disposition: the biological fate of chemicals, 38(9), 1532-1537 (2010-06-15)
Phenacetin was withdrawn from the market because it caused renal failure in some patients. Many reports indicated that the nephrotoxicity of phenacetin is associated with the hydrolyzed metabolite, p-phenetidine. Acetaminophen (APAP), the major metabolite of phenacetin, is also hydrolyzed to
Tatsuki Fukami et al.
Drug metabolism and pharmacokinetics, 27(5), 466-477 (2012-07-21)
In this review, novel aspects of the role of esterases, which contribute to the metabolism of 10% of therapeutic drugs, are described. Esterases hydrolyze the compounds that contain ester, amide, and thioester bonds, which cause prodrug activation or detoxification. Among

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service