Skip to Content
Merck
All Photos(2)

Key Documents

AV44910

Sigma-Aldrich

Anti-MOSPD3 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-CDS3, Anti-Motile sperm domain containing 3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$547.00

$547.00


Estimated to ship on14 May 2025



Select a Size

Change View
100 μL
$547.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$547.00


Estimated to ship on14 May 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

24 kDa

species reactivity

mouse, human, rat

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... MOSPD3(64598)

Immunogen

Synthetic peptide directed towards the C terminal region of human MOSPD3

Application

Anti-MOSPD3 antibody produced in rabbit is suitable for western blotting at a concentration of 5.0μg/ml and for immunohistochemistry of paraffin-embedded tissue sections at a concentration of 4-8μg/ml.

Biochem/physiol Actions

Motile sperm domain containing 3 (MOSPD3) is a multi-pass membrane protein containing a major sperm protein (MSP) domain. It has a crucial role in the development of right ventricle. Mutations in the gene encoding for MOSPD3 result in defective cardiac development and neonatal lethality.[1]

Sequence

Synthetic peptide located within the following region: FLLFLLTGIVSVAFLLLPLPDELGSQLPQVLHVSLGQKLVAAYVLGLLTM

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gurman S Pall et al.
Genomics, 84(6), 1051-1059 (2004-11-10)
We have identified and characterized a gene, Mospd3 on mouse chromosome 5 using gene trapping in ES cells. MOSPD3 is part of a family of proteins, including MOSPD1, which is defined by the presence of a major sperm protein (MSP)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service