Skip to Content
Merck
All Photos(3)

Key Documents

AV44817

Sigma-Aldrich

Anti-RHOT1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-ARHT1, Anti-FLJ11040, Anti-FLJ12633, Anti-MIRO-1, Anti-Ras homolog gene family, member T1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$716.00

$716.00


Check Cart for Availability


Select a Size

Change View
100 μL
$716.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$716.00


Check Cart for Availability

antibody form

affinity isolated antibody

Quality Level

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

71 kDa

species reactivity

rabbit, horse, bovine, rat, human, mouse, guinea pig, dog

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RHOT1(55288)

General description

Anti-RHOT1 polyclonal antibody reacts with RHOT1 in chicken, bovine, human, zebrafish, pig, rat, canine, and mouse.
Ras homolog gene family, member T1 (RHOT1, MIRO-1) is an atypical Rho GTPase involved in calcium sensitive mitochondrial trafficking. Miro interacts with the kinesin-binding proteins GRIF-1 and OIP106 forming a link between the mitochondria and the trafficking apparatus of the microtubules. Miro1 and the kinesin adaptor Grif-1 play an important role in regulating mitochondrial transport in neurons. Miro1 is a calcium sensor for glutamate receptor-dependent localization of mitochondria at synapses.

Immunogen

Synthetic peptide directed towards the N terminal region of human RHOT1

Application

Anti-RHOT1 polyclonal antibody is suitable for use in western blotting and immunohistochemical (IHC) techniques. The antibody is used as a probe to determine the role of RHOT1 in calcium-sensitive mitochondrial trafficking.

Biochem/physiol Actions

Mitochondrial GTPase involved in mitochondrial trafficking. RHOT1 is probably involved in control of anterograde transport of mitochondria and their subcellular distribution.

Sequence

Synthetic peptide located within the following region: MKKDVRILLVGEPRVGKTSLIMSLVSEEFPEEVPPRAEEITIPADVTPER

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ji Geng et al.
Developmental cell, 58(7), 597-615 (2023-04-12)
Loss of fragile X messenger ribonucleoprotein (FMRP) causes fragile X syndrome (FXS), the most prevalent form of inherited intellectual disability. Here, we show that FMRP interacts with the voltage-dependent anion channel (VDAC) to regulate the formation and function of endoplasmic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service