Skip to Content
Merck
All Photos(6)

Key Documents

AV44142

Sigma-Aldrich

Anti-SLC39A5 (AB1) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-LZT-Hs7, Anti-MGC34778, Anti-Solute carrier family 39 (metal ion transporter), member 5, Anti-ZIP5

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$609.00

$609.00


Estimated to ship on17 April 2025



Select a Size

Change View
100 μL
$609.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$609.00


Estimated to ship on17 April 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

56 kDa

species reactivity

mouse, human, rat, rabbit

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Immunogen

Synthetic peptide directed towards the N terminal region of human SLC39A5

Application

Anti-SLC39A5 (AB1) antibody produced in rabbit is suitable for western blotting at a concentration of 0.5μg/ml and for immunohistochemistry of paraffin-embedded tissue sections at a concentration of 4-8μg/ml.

Biochem/physiol Actions

SLC39A5 (ZIP5) is a zinc uptake transporter that specifically transports Zn+2 ions. It is expressed in intestine, pancreas, liver and kidney. ZIP5 maintains zinc homeostasis and plays an important role in the uptake of dietary zinc across apical membrane of enterocytes in the intestine. Zinc transport by ZIP5 is important to protect the pancreatic acinar cells against zinc toxicity.

Sequence

Synthetic peptide located within the following region: MGSPVSHLLAGFCVWVVLGWVGGSVPNLGPAEQEQNHYLAQLFGLYGENG

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jim Geiser et al.
PloS one, 8(11), e82149-e82149 (2013-12-05)
ZIP5 localizes to the baso-lateral membranes of intestinal enterocytes and pancreatic acinar cells and is internalized and degraded coordinately in these cell-types during periods of dietary zinc deficiency. These cell-types are thought to control zinc excretion from the body. The
Benjamin P Weaver et al.
Biometals : an international journal on the role of metal ions in biology, biochemistry, and medicine, 25(2), 319-335 (2011-11-25)
Translation of the basolateral zinc transporter ZIP5 is repressed during zinc deficiency but Zip5 mRNA remains associated with polysomes and can be rapidly translated when zinc is repleted. Herein, we examined the mechanisms regulating translation of Zip5. The 3'-untranslated region
Fudi Wang et al.
The Journal of biological chemistry, 279(49), 51433-51441 (2004-08-24)
The mouse and human Zip5 proteins are members of the ZIP family of metal ion transporters. In this study, we present evidence that mouse Zip5 is a zinc uptake transporter that is specific for Zn(II) over other potential metal ion

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service