Skip to Content
Merck
All Photos(6)

Key Documents

AV43931

Sigma-Aldrich

Anti-SLC39A6 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-LIV-1, Anti-Solute carrier family 39 (Zinc transporter), member 6

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$554.00

$554.00


Estimated to ship on24 May 2025



Select a Size

Change View
100 μL
$554.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$554.00


Estimated to ship on24 May 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

48 kDa

species reactivity

human, mouse, rat, sheep, horse, dog, bovine, rabbit

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC39A6(25800)

General description

Solute carrier family 39 (Zinc transporter), member 6/zinc transporter 6 (SLC39A6, LIV1, ZIP6) is a mediator of zinc (Zn2+) transport and intracellular zinc homeostasis. SLC39A6/LIV1 is involves in important processes such as epithelial-to-mesenchymal transition (EMT) in human pancreatic, breast, and prostate cancer cells. SLC39A6/LIV1 has been shown to be a critical mediator responsible for HDACi-induced apoptosis.

Specificity

Anti-SLC39A6 polyclonal antibody reacts with bovine, canine, human, mouse, and rat solute carrier family 39 (Zinc transporter), member 6 proteins.

Immunogen

Synthetic peptide directed towards the middle region of human SLC39A6

Application

Anti-SLC39A6 polyclonal antibody is used to tag solute carrier family 39 (Zinc transporter), member 6/zinc transporter 6 for detection and quantitation by Western blotting and in plasma by immunohistochemical (IHC) techniques. It is used as a probe to determine the roles of solute carrier family 39 (Zinc transporter), member 6/zinc transporter 6 in intracellular zinc homeostasis, epithelial-to-mesenchymal transition (EMT) in cancer cells, and in HDACi-induced apoptosis.

Biochem/physiol Actions

Zinc is an essential cofactor for hundreds of enzymes. It is involved in protein, nucleic acid, carbohydrate, and lipid metabolism, as well as in the control of gene transcription, growth, development, and differentiation. SLC39A6 belongs to a subfamily of proteins that show structural characteristics of zinc transporters.

Sequence

Synthetic peptide located within the following region: RSCLIHTSEKKAEIPPKTYSLQIAWVGGFIAISIISFLSLLGVILVPLMN

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service