Skip to Content
Merck
All Photos(3)

Key Documents

AV43158

Sigma-Aldrich

Anti-PRPF19 (AB2) antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-NMP200, Anti-PRP19, Anti-PRP19/PSO4 pre-mRNA processing factor 19 homolog (S. cerevisiae), Anti-PSO4, Anti-UBOX4, Anti-hPSO4

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$547.00

$547.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1068


Select a Size

Change View
100 μL
$547.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$547.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1068

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

55 kDa

species reactivity

guinea pig, mouse, dog, rabbit, rat, human, horse, bovine

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PRPF19(27339)

Immunogen

Synthetic peptide directed towards the middle region of human PRPF19

Biochem/physiol Actions

PRPF19 plays a role in DNA double-strand break (DSB) repair and pre-mRNA splicing reaction. It binds double-stranded DNA in a sequence-nonspecific manner. PRPF19 acts as a structural component of the nuclear framework. It may also serve as a support for spliceosome binding and activity. It is essential for spliceosome assembly in a oligomerization-dependent manner and might also be important for spliceosome stability. It also may have E3 ubiquitin ligase activity. The PSO4 complex is required in the DNA interstrand cross-links (ICLs) repair process. Overexpression of PRPF19 might extend the cellular life span by increasing the resistance to stress or by improving the DNA repair capacity of the cells.In S. cerevisiae, Pso4 has pleiotropic functions in DNA recombination and in error-prone nonhomologous end-joining DNA repair.[supplied by OMIM].

Sequence

Synthetic peptide located within the following region: PSVVGAGEPMDLGELVGMTPEIIQKLQDKATVLTTERKKRGKTVPEELVK

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service