Skip to Content
Merck
All Photos(3)

Key Documents

AV43006

Sigma-Aldrich

Anti-UBE2D1 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-E2(17)KB1, Anti-SFT, Anti-UBC4/5, Anti-UBCH5, Anti-UBCH5A, Anti-Ubiquitin-conjugating enzyme E2D 1 (UBC4/5 homolog, yeast)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$547.00

$547.00


Estimated to ship on21 May 2025



Select a Size

Change View
100 μL
$547.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$547.00


Estimated to ship on21 May 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

16 kDa

species reactivity

horse, bovine, pig, goat, rat, dog, mouse, human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... UBE2D1(7321)

Immunogen

Synthetic peptide directed towards the middle region of human UBE2D1

Biochem/physiol Actions

The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. UBE2D1 is a member of the E2 ubiquitin-conjugating enzyme family. This enzyme is closely related to a stimulator of iron transport (SFT), and is up-regulated in hereditary hemochromatosis. It also functions in the ubiquitination of the tumor-suppressor protein p53 and the hypoxia-inducible transcription factor HIF1alpha by interacting with the E1 ubiquitin-activating enzyme and the E3 ubiquitin-protein ligases.

Sequence

Synthetic peptide located within the following region: SQWSPALTVSKVLLSICSLLCDPNPDDPLVPDIAQIYKSDKEKYNRHARE

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu Li et al.
Cancer management and research, 13, 7543-7557 (2021-10-12)
Cisplatin (DDP) treatment is one of the most predominant chemotherapeutic strategies for patients with gastric cancer (GC). LncRNA noncoding RNA HLA complex group 11 (lncRNA HCG11) has been confirmed to promote GC progression. This study attempted to investigate the underlying

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service