Skip to Content
Merck
All Photos(3)

Key Documents

AV42475

Sigma-Aldrich

Anti-PLUNC antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-LUNX, Anti-NASG, Anti-Palate, lung and nasal epithelium carcinoma associated, Anti-SPLUNC1, Anti-SPURT, Anti-bA49G10.5

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$609.00

$609.00


Check Cart for Availability


Select a Size

Change View
100 μL
$609.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$609.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

28 kDa

species reactivity

guinea pig, goat, rat, bovine, human, dog, mouse

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PLUNC(51297)

General description

PLUNC-like proteins display sequence homology with BPI (bactericidal/permeability-increasing protein) is a 456-residue cationic protein shown to possess both bactericidal and LPS (lipopolysaccharide)-binding activities. Palate, lung and nasal epithelium carcinoma associated/secretory protein in upper respiratory tracts (PLUNC, LUNX, NASG, SPURT, SPLUNC1) appears to support antimicrobial and anti-inflammatory functions in Gram-negative bacteria-induced respiratory infection.

Specificity

Anti-PLUNC polyclonal antibody reacts with human, mouse, rat, pig, canine, and bovine PLUNC1 proteins.

Immunogen

Synthetic peptide directed towards the middle region of human PLUNC

Application

Anti-PLUNC polyclonal antibody is used to tag palate, lung and nasal epithelium carcinoma associated/secretory protein in upper respiratory tracts (PLUNC1) for detection and quantitation by Western blotting and in plasma by immunohistochemical (IHC) techniques. It is used as a probe to determine the roles of palate, lung and nasal epithelium carcinoma associated/secretory protein in upper respiratory tracts in defense against Gram-negative bacterial-induced respiratory infection.

Biochem/physiol Actions

PLUNC is the human homolog of murine plunc and is specifically expressed in the upper airways and nasopharyngeal regions. The exact biological function of this protein is not known, however, it has been suggested to be involved in inflammatory responses to irritants in the upper airways. It may also serve as a potential molecular marker for detection of micrometastasis in non-small-cell lung cancer.This gene is the human homolog of murine plunc, and like the mouse gene, is specifically expressed in the upper airways and nasopharyngeal regions. The exact biological function of this gene is not known, however, it has been suggested to be involved in inflammatory responses to irritants in the upper airways. It may also serve as a potential molecular marker for detection of micrometastasis in non-small-cell lung cancer. Multiple transcript variants resulting from alternative splicing in the 3′ UTR have been detected, but the full-length nature of only two is known.

Sequence

Synthetic peptide located within the following region: GLNNIIDIKVTDPQLLELGLVQSPDGHRLYVTIPLGIKLQVNTPLVGASL

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrew K Beppu et al.
Nature communications, 14(1), 5814-5814 (2023-09-20)
Epithelial plasticity has been suggested in lungs of mice following genetic depletion of stem cells but is of unknown physiological relevance. Viral infection and chronic lung disease share similar pathological features of stem cell loss in alveoli, basal cell (BC)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service