Skip to Content
Merck
All Photos(1)

Key Documents

AV42318

Sigma-Aldrich

Anti-SLC20A2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-GLVR2, Anti-Glvr-2, Anti-MLVAR, Anti-PIT-2, Anti-Solute carrier family 20 (phosphate transporter), member 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$609.00

$609.00


Check Cart for Availability


Select a Size

Change View
100 μL
$609.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$609.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

70 kDa

species reactivity

guinea pig, bovine, rat, human, dog, mouse, horse, rabbit

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC20A2(6575)

General description

Solute carrier family 20 (phosphate transporter), member 2 (SLC20A2, GLVR2, MLVAR, PIT-2) is a type III Na+-dependent Pi transporter responsible for the transport of inorganic phosphate (Pi) and maintenance of Pi homeostasis that supports biological processes such as nucleic acid synthesis, tooth mineralization, skeletal development and various signaling cascades. Defective SLC20a2 is associated with idiopathic basal ganglia calcification (Fahr′s syndrome).

Specificity

Anti-SLC20A2 polyclonal antibody reacts with bovine, chicken, human, mouse, rat, canine, and zebrafish solute carrier family 20 (phosphate transporter) member 2 proteins.

Immunogen

Synthetic peptide directed towards the N terminal region of human SLC20A2

Application

Anti-SLC20A2 polyclonal antibody is used to tag solute carrier family 20 (phosphate transporter) member 2/ gibbon ape leukemia virus 2 protein for detection and quantitation by Western blotting and in plasma by immunohistochemical (IHC) techniques. It is used as a probe to determine the roles of solute carrier family 20 (phosphate transporter) member 2/gibbon ape leukemia virus 2 protein in Pi homeostasis.

Sequence

Synthetic peptide located within the following region: DVNLYNETVETLMAGEVSAMVGSAVWQLIASFLRLPISGTHCIVGSTIGF

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service