Skip to Content
Merck
All Photos(4)

Key Documents

AV41474

Sigma-Aldrich

Anti-BHMT (AB1) antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-βSerine-homocysteine methyltransferase

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$547.00

$547.00


Estimated to ship on13 April 2025



Select a Size

Change View
100 μL
$547.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$547.00


Estimated to ship on13 April 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

45 kDa

species reactivity

rabbit, guinea pig, mouse, horse, human, rat, dog, bovine

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BHMT(635)

Immunogen

Synthetic peptide directed towards the N terminal region of human BHMT

Biochem/physiol Actions

BHMT is a cytosolic enzyme that catalyzes the conversion of betaine and homocysteine to dimethylglycine and methionine, respectively. Defects in its gene could lead to hyperhomocyst(e)inemia, but such a defect has not yet been observed.Betaine-homocysteine methyltransferase is a cytosolic enzyme that catalyzes the conversion of betaine and homocysteine to dimethylglycine and methionine, respectively. Defects in BHMT could lead to hyperhomocyst(e)inemia,but such a defect has not yet been observed.

Sequence

Synthetic peptide located within the following region: AVEHPEAVRQLHREFLRAGSNVMQTFTFYASEDKLENRGNYVLEKISGQE

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y F Zhou et al.
Journal of dairy science, 101(12), 11384-11395 (2018-10-15)
Insufficient supply of Met and choline (Chol) around parturition could compromise hepatic metabolism and milk protein synthesis in dairy cows. Mechanistic responses associated with supply of Met or Chol in primary liver cells enriched with hepatocytes (PHEP) from cows have
Anders Øverbye et al.
Autophagy, 7(9), 1011-1027 (2011-05-26)
To investigate the stepwise autophagic-lysosomal processing of hepatocellular proteins, the abundant cytosolic enzyme, betaine:homocysteine methyltransferase (BHMT) was used as a probe. Full-length (45 kDa) endogenous BHMT was found to be cleaved in an autophagy-dependent (3-methyladenine-sensitive) manner in isolated rat hepatocytes

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service