Skip to Content
Merck
All Photos(2)

Key Documents

AV41198

Sigma-Aldrich

Anti-DND1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Dead end homolog 1 (zebrafish), Anti-MGC34750, Anti-RBMS4

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$609.00

$609.00


Estimated to ship on27 May 2025



Select a Size

Change View
100 μL
$609.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$609.00


Estimated to ship on27 May 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

39 kDa

species reactivity

rat, mouse, human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DND1(373863)

General description

Dead end homolog 1 (zebrafish) is an RNA binding protein found in primordial germ cells during embryogenesis. DND1 is essential for germ cell survival. Dnd1 appears to have a critical role in germ-cell and germ cell-tumor development. Dnd1 acts, at least in part, by protecting certain mRNAs from micro-RNA (miRNA)-mediated repression.

Specificity

Anti-DND1 polyclonal antibody reacts with human, mouse, rat, bovine, and canine dead end homolog 1 proteins.

Immunogen

Synthetic peptide directed towards the C terminal region of human DND1

Application

Anti-DND1 polyclonal antibody is used to tag dead end homolog 1 proteins for detection and quantitation by Western blotting and in cells and tissues by immunohistochemical (IHC) techniques. It is used as a probe to study the role of dead end homolog 1 proteins in germ-cell and germ cell-tumor development, especially at the level of miRNA mediated repression of mRNA transcription.

Biochem/physiol Actions

DND1 contains 2 RRM (RNA recognition motif) domains. It may play a role during primordial germ cell (PGC) development. However, DND1 does not seem to be essential for PGC migration

Sequence

Synthetic peptide located within the following region: HRFWYQVVIPGHPVPFSGLIWVVLTLDGRDGHEVAKDAVSVRLLQALSES

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marta Blanes-García et al.
Fish physiology and biochemistry (2024-04-19)
Identification of specific molecular markers for spermatogonial stem cells in teleost is crucial for enhancing the efficacy of reproductive biotechnologies in aquaculture, such as transplantation and surrogate production in fishes. Since it is not yet possible to distinguish spermatogonial stem

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service