Skip to Content
Merck
All Photos(1)

Key Documents

AV41187

Sigma-Aldrich

Anti-CPEB2 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Cytoplasmic polyadenylation element binding protein 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

37 kDa

species reactivity

dog, guinea pig, rat, rabbit, bovine, mouse, human, horse

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CPEB2(132864)

General description

Cytoplasmic polyadenylation element binding protein 2 (CPEB2) is an RNA binding protein that regulates cytoplasmic polyadenylation and translation of mRNAs in germ cells and neural cells. CPEB2 is involved in the regulation of the oxygen homoeostasis transcriptional master regulator gene hypoxia-inducible factor-1 (HIF-1).

Specificity

Anti-CPEB2 polyclonal antibody reacts with bovine, chicken, canine, pig, human, mouse, rat, and zebrafish cytoplasmic polyadenylation element binding protein 2 proteins.

Immunogen

Synthetic peptide directed towards the middle region of human CPEB2

Application

Anti-CPEB2 polyclonal antibody is used to tag cytoplasmic polyadenylation element binding protein 2 proteins for detection and quantitation by Western blotting and in cells and tissues by immunohistochemical (IHC) techniques. It is used as a probe to study the role of CPEB2 in the regulation of mRNA transcription in germ cells and neural cells.

Biochem/physiol Actions

CPEB2 is highly similar to cytoplasmic polyadenylation element binding protein (CPEB), an mRNA-binding protein that regulates cytoplasmic polyadenylation of mRNA as a trans factor in oogenesis and spermatogenesis. Studies of the similar gene in mice suggested a possible role of this protein in transcriptionally inactive haploid spermatids.

Sequence

Synthetic peptide located within the following region: DTDPELKYPKGAGRVAFSNQQSYIAAISARFVQLQHGDIDKRVEVKPYVL

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service