Skip to Content
Merck
All Photos(3)

Key Documents

AV40071

Sigma-Aldrich

Anti-RNF8 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-FLJ12013, Anti-KIAA0646, Anti-Ring finger protein 8

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$547.00

$547.00


Check Cart for Availability


Select a Size

Change View
100 μL
$547.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$547.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

55 kDa

species reactivity

pig, mouse, bovine, rat, human, horse

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RNF8(9025)

General description

Ring finger protein 8 (RNF8) is an E3 ubiquitin ligase which ubiquitinates proteins for degradation by proteosomes. RNF8 is involved in DNA double-strand break (DSB) repair, wherein RNF8 and RNF168 control the recruitment of 53BP1 to DNA damage sites and ubiquitination of histone-2A. RNF8 also mediates chromatin decondensation to create a local chromatin environment that is permissive to the assembly of checkpoint and repair machineries at DNA lesions.

Specificity

Anti-RNF8 polyclonal antibody reacts with human, mouse, rat, and bovine ring finger protein 8 proteins.

Immunogen

Synthetic peptide directed towards the C terminal region of human RNF8

Application

Anti-RNF8 polyclonal antibody is used to tag ring finger protein 8 for detection and quantitation by Western blotting and in plasma by immunohistochemical (IHC) techniques. It is used as a probe to determine the roles ring finger protein 8 in processes such as DNA double-strand break (DSB) repair.

Biochem/physiol Actions

RNF8 contains a RING finger motif and a FHA domain. This protein has been shown to interact with several class II ubiquitin-conjugating enzymes (E2), including UBE2E1/UBCH6, UBE2E2, and UBE2E3, and may act as an ubiquitin ligase (E3) in the ubiquitination of certain nuclear proteins.The protein encoded by this gene contains a RING finger motif and a FHA domain. This protein has been shown to interact with several class II ubiquitin-conjugating enzymes (E2), including UBE2E1/UBCH6, UBE2E2, and UBE2E3, and may act as an ubiquitin ligase (E3) in the ubiquitination of certain nuclear proteins. Alternatively spliced transcript variants encoding distinct isoforms have been reported.

Sequence

Synthetic peptide located within the following region: MEELNRSKKDFEAIIQAKNKELEQTKEEKEKMQAQKEEVLSHMNDVLENE

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Dafne Cardamone et al.
Cell reports, 8(1), 163-176 (2014-06-24)
Timely and selective recruitment of transcription factors to their appropriate DNA-binding sites represents a critical step in regulating gene activation; however, the regulatory strategies underlying each factor's effective recruitment to specific promoter and/or enhancer regions are not fully understood. Here

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service