Skip to Content
Merck
All Photos(1)

Key Documents

AV39504

Sigma-Aldrich

Anti-PRDM12 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-PR domain containing 12

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$547.00

$547.00


Check Cart for Availability


Select a Size

Change View
100 μL
$547.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$547.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

40 kDa

species reactivity

rat, human, mouse

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PRDM12(59335)

General description

PR domain proteins (PRDM) are a small family of kruppel-like zinc finger transcription factors involved in cell differentiation and tumorigenesis. PR domain containing 12 (PRDM12) is a putative tumor suppressor that may be implicated in poor outcome chronic myeloid leukemia (CML).

Specificity

Anti-PRDM12 polyclonal antibody reacts with zebrafish, human, mouse, rat, and bovine PR domain containing 12 proteins.

Immunogen

Synthetic peptide directed towards the C terminal region of human PRDM12

Application

Anti-PRDM12 polyclonal antibody is used to tag PR domain containing 12 protein for detection and quantitation by Western blotting and in plasma by immunohistochemical (IHC) techniques. It is used as a probe to determine the roles of PR domain containing 12 protein in cell differentiation and tumorigenesis such as chronic myeloid leukemia.

Biochem/physiol Actions

PRDM12 belongs to PR-domain family which are involved in human cancers in an unusual yin-yang fashion. Two products are normally produced from a PR-domain family member which differ by the presence or absence of the PR domain; the PR-plus product is disrupted or underexpressed whereas the PR-minus product is present or overexpressed in cancer cells. This imbalance in the amount of the two products, a result of either genetic or epigenetic events, appears to be an important cause of malignancy. PRDM12 may have a potential role in the pathogenesis of chronic myeloid leukaemia with derivative chromosome 9 deletion.

Sequence

Synthetic peptide located within the following region: NHVRLHTGERPYKCQVCQSAYSQLAGLRAHQKSARHRPPSTALQAHSPAL

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service