Skip to Content
Merck
All Photos(1)

Key Documents

AV39337

Sigma-Aldrich

Anti-ZFP64 (AB1) antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Zinc finger protein 64 homolog (mouse)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$482.00

$482.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1199


Select a Size

Change View
100 μL
$482.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$482.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1199

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

69 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ZFP64(55734)

General description

ZFP64 is a zinc-finger protein that enhances p65 stimulation and thereby promotes TLR-induced production of pro-inflammatory cytokines and type I interferons in macrophages.
Rabbit Anti-ZFP64 antibody recognizes bovine, canine, human, and pig ZFP64.

Immunogen

Synthetic peptide directed towards the N terminal region of human ZFP64

Application

Rabbit Anti-ZFP64 antibody is suitable for western blotting applications at a concentration of 2.5 μg/ml.

Biochem/physiol Actions

ZFP64 belongs to the krueppel C2H2-type zinc-finger protein family. It contains 9 C2H2-type zinc fingers and may be involved in transcriptional regulation.

Sequence

Synthetic peptide located within the following region: MNASSEGESFAGSVQIPGGTTVLVELTPDIHICGICKQQFNNLDAFVAHK

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chunmei Wang et al.
The Journal of biological chemistry, 288(34), 24600-24608 (2013-07-17)
The molecular mechanisms that fine-tune the Toll-like receptor (TLR)-triggered innate immune response need further investigation. As an important transcription factor, zinc finger proteins (ZFPs) play important roles in many cell functions, including development, differentiation, tumorigenesis, and functions of the immune

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service