Skip to Content
Merck
All Photos(1)

Documents

AV38799

Sigma-Aldrich

Anti-TRIM16 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Tripartite motif-containing 16

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

64 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TRIM16(10626)

Immunogen

Synthetic peptide directed towards the N terminal region of human TRIM16

Biochem/physiol Actions

Tripartite motif containing 16 (TRIM16) is transcriptional repressor belonging to tripartite motif family. It is described as a tumor suppressor protein that interacts with vimentin and E2F1 proteins. It inhibits cell proliferation by inducing cell cycle arrest in G1/S phase in neuroblastoma cells. TRIM16 has the ability to heterodimerize with other TRIM proteins and exhibits E3 ubiquitin ligase activity.

Sequence

Synthetic peptide located within the following region: PLPRATAQPPAPLSPDSGSPSPDSGSASPVEEEDVGSSEKLGRETEEQDS

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jessica L Bell et al.
Cell cycle (Georgetown, Tex.), 12(6), 889-898 (2013-02-21)
Neuroblastoma is the most common solid tumor in childhood and represents 15% of all children's cancer deaths. We have previously demonstrated that tripartite motif 16 (TRIM16), a member of the RING B-box coiled-coil (RBCC)/tripartite totif (TRIM) protein family, has significant
Patrick Y Kim et al.
Apoptosis : an international journal on programmed cell death, 18(5), 639-651 (2013-02-14)
TRIM16 exhibits tumour suppressor functions by interacting with cytoplasmic vimentin and nuclear E2F1 proteins in neuroblastoma and squamous cell carcinoma cells, reducing cell migration and replication. Reduced TRIM16 expression in a range of human primary malignant tissues correlates with increased
Jessica L Bell et al.
PloS one, 7(5), e37470-e37470 (2012-05-26)
The TRIM family of proteins is distinguished by its tripartite motif (TRIM). Typically, TRIM proteins contain a RING finger domain, one or two B-box domains, a coiled-coil domain and the more variable C-terminal domains. TRIM16 does not have a RING

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service