Skip to Content
Merck
All Photos(2)

Key Documents

AV38613

Sigma-Aldrich

Anti-NFIL3 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Nuclear factor, interleukin 3 regulated

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$482.00

$482.00


Check Cart for Availability


Select a Size

Change View
100 μL
$482.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$482.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

51 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NFIL3(4783)

General description

Nuclear factor, interleukin 3 regulated (NFIL3, E4BP4) is a transcription factor involved in development of haematopoietic lineages. NFIL3/E4BP4 is required for development of NK cells and CD8α(+) conventional dendritic cells, and myeloid lineage commitment. NFIL3/E4BP4 also facilitates macrophage activation, polarisation of CD4(+) T cell responses and B cell class switching to IgE.

Specificity

Anti-NFIL3 polyclonal antibody reacts with human, bovine, and canine nuclear factor, interleukin 3 regulated proteins.

Immunogen

Synthetic peptide directed towards the N terminal region of human NFIL3

Application

Anti-NFIL3 polyclonal antibody is used to tag nuclear factor, interleukin 3 regulated for detection and quantitation by Western blotting and in plasma by immunohistochemical (IHC) techniques. It is used as a probe to determine the roles of nuclear factor, interleukin 3 regulated in haematopoietic lineages development and specialized immune function regulation.

Biochem/physiol Actions

NFIL3 is a basic leucine zipper (bZIP) transcription factor that represses or activates transcription in non-osteoblastic cells. NFIL3 play a role in attenuation of PTH target gene transcription in osteoblasts. NFIL3 repression domain interacts specifically with the TBP binding repressor protein Dr1.Glucocorticoid-mediated upregulation of the bZIP transcriptional repressor gene, NFIL3, is dependent on [Ca2+]i levels, and correlates with Glucocorticoid-evoked apoptosis of Glucocorticoid-sensitive CEM-C7-14 cells. NFIL3 is involved in a distinct growth factor-regulated signaling pathway that is responsible for the survival of early B-cell progenitors, and whose alteration by E2A-HLF leads to childhood B lineage leukemia.

Sequence

Synthetic peptide located within the following region: MQLRKMQTVKKEQASLDASSNVDKMMVLNSALTEVSEDSTTGEDVLLSEG

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service