Skip to Content
Merck
All Photos(3)

Key Documents

AV35123

Sigma-Aldrich

Anti-VDAC2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-FLJ23841, Anti-RP11-375G3.1, Anti-Voltage-dependent anion channel 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$537.00

$537.00


Estimated to ship on02 June 2025



Select a Size

Change View
100 μL
$537.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$537.00


Estimated to ship on02 June 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

32 kDa

species reactivity

dog, bovine, guinea pig, rabbit, rat, human, horse, mouse

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... VDAC2(7417)

General description

VDAC2 is a voltage-dependant anion channel that forms a pathway for metabolite diffusion across the outer membrane of mitochondria. Studies have reported that VDAC2 interacts with BAK and subsequently regulates mitochondrial apoptosis/cell death[1][2].
Rabbit Anti-VDAC2 antibody recognizes chicken, human, mouse, rat, bovine, pig, canine, rabbit, and zebrafish VDAC2.

Immunogen

Synthetic peptide directed towards the N terminal region of human VDAC2

Application

Rabbit Anti-VDAC2 antibody is suitable for western blot applications at a concentration of 0.25 μg/ml.

Biochem/physiol Actions

VDAC2 forms a channel through the mitochondrial outer membrane that allows diffusion of small hydrophilic molecules. The channel adopts an open conformation at low or zero membrane potential and a closed conformation at potentials above 30-40 mV. The open state has a weak anion selectivity whereas the closed state is cation-selective.

Sequence

Synthetic peptide located within the following region: MATHGQTCARPMCIPPSYADLGKAARDIFNKGFGFGLVKLDVKTKSCSGV

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael Lazarou et al.
The Journal of biological chemistry, 285(47), 36876-36883 (2010-09-21)
Bax and Bak are pro-apoptotic factors that are required for cell death by the mitochondrial or intrinsic pathway. Bax is found in an inactive state in the cytosol and upon activation is targeted to the mitochondrial outer membrane where it
Soumya Sinha Roy et al.
EMBO reports, 10(12), 1341-1347 (2009-10-13)
Truncated BID (tBID), a proapoptotic BCL2 family protein, induces BAK/BAX-dependent release of cytochrome c and other mitochondrial intermembrane proteins to the cytosol to induce apoptosis. The voltage-dependent anion channels (VDACs) are the primary gates for solutes across the outer mitochondrial
Mayumi Watanabe et al.
Toxicology, 322, 43-50 (2014-05-08)
Parkin is an E3 ubiquitin ligase involved in the elimination of damaged mitochondria. Ubiquitination of mitochondrial substrates by Parkin results in proteasomal as well as lysosomal degradation of mitochondria, the latter of which is executed by the autophagy machinery and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service