Skip to Content
Merck
All Photos(1)

Key Documents

AV34917

Sigma-Aldrich

Anti-CLCNKB (AB1) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Chloride channel Kb

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$716.00

$716.00


Estimated to ship on26 May 2025



Select a Size

Change View
100 μL
$716.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$716.00


Estimated to ship on26 May 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

76 kDa

species reactivity

pig, horse, bovine, human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CLCNKB(1188)

General description

CLCNKB is a voltage-gated chloride channel that may modulate reabsorption of renal salts. Genetic variations in CLCNKB have been implicated in Gitelman and Bartter syndromes, as well as in the induction of ClC-Kb chloride channel functions.
Rabbit Anti-CLCNKB antibody recognizes human, mouse, rat, canine, rabbit, and bovine CLCNKB.

Immunogen

Synthetic peptide directed towards the C terminal region of human CLCNKB

Application

Rabbit Anti-CLCNKB antibody is suitable for western blot applications at a concentration of 0.5 μg/ml.

Biochem/physiol Actions

Chloride channel Kb (CLCNKB) is a member of the CLC family of voltage-gated chloride channels, which comprises at least 9 mammalian chloride channels. Each is believed to have 12 transmembrane domains and intracellular N and C termini. Mutations in CLCNKB result in the autosomal recessive Type III Bartter Syndrome. CLCNKA and CLCNKB are closely related (94% sequence identity), tightly linked (separated by 11 kb of genomic sequence) and are both expressed in mammalian kidney.

Sequence

Synthetic peptide located within the following region: ILAAGCPTEPVTLKLSPETSLHEAHNLFELLNLHSLFVTSRGRAVGCVSW

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nikola Jeck et al.
Kidney international, 65(1), 190-197 (2003-12-17)
Tubular transepithelial reabsorption of chloride along the nephron is a major determinant of body salt and water homeostasis and blood pressure regulation. About 40% of the glomerulary filtered sodium chloride are reabsorbed in the distal nephrons. Vectorial transepithelial sodium chloride
Israel Zelikovic et al.
Kidney international, 63(1), 24-32 (2002-12-11)
Gitelman syndrome (GS) and Bartter syndrome (BS) are hereditary hypokalemic tubulopathies with distinct phenotypic features. GS has been considered a genetically homogeneous disorder caused by mutation in the gene encoding the NaCl cotransporter (TSC) of the distal convoluted tubule. In

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service