Skip to Content
Merck
All Photos(2)

Key Documents

AV34773

Sigma-Aldrich

Anti-HOXA1 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Homeobox A1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$482.00

$482.00


Check Cart for Availability


Select a Size

Change View
100 μL
$482.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$482.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

14 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HOXA1(3198)

General description

HOXA1 is a homeobox transcription factor that may regulate differentiation and morphogenesis in cells. Mutations in HOXA1 have been linked to the risk of autism spectrum disorders and defects in the central nervous system.
Rabbit HOXA1 antibody recognizes human, mouse, rat, rabbit, bovine, and canine HOXA1.

Immunogen

Synthetic peptide directed towards the middle region of human HOXA1

Application

Rabbit HOXA1 antibody is suitable for western blot (2.5 μg/ml) and IHC (4-8 μg/ml) applications.

Biochem/physiol Actions

In vertebrates, the genes encoding the class of transcription factors called homeobox genes are found in clusters named A, B, C, and D on four separate chromosomes. Expression of these proteins is spatially and temporally regulated during embryonic development. HOXA1 is part of the A cluster on chromosome 7 and encodes a DNA-binding transcription factor which may regulate gene expression, morphogenesis, and differentiation. The encoded protein may be involved in the placement of hindbrain segments in the proper location along the anterior-posterior axis during development.

Sequence

Synthetic peptide located within the following region: NLGVSYSHSSCGPSYGSQNFSAPYSPYALNQEADPPRSLSLPRIGDIFSS

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J L Ingram et al.
Teratology, 62(6), 393-405 (2000-11-25)
Family studies have demonstrated that the autism spectrum disorders (ASDs) have a major genetic etiologic component, but expression and penetrance of the phenotype are variable. Mice with null mutations of Hoxa1 or Hoxb1, two genes critical to hindbrain development, have
M Rossel et al.
Development (Cambridge, England), 126(22), 5027-5040 (1999-10-26)
The analysis of mice mutant for both Hoxa1 and Hoxb1 suggests that these two genes function together to pattern the hindbrain. Separately, mutations in Hoxa1 and Hoxb1 have profoundly different effects on hindbrain development. Hoxa1 mutations disrupt the rhombomeric organization

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service