Skip to Content
Merck
All Photos(1)

Key Documents

AV34420

Sigma-Aldrich

Anti-HEXIM1 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Hexamethylene bis-acetamide inducible 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$482.00

$482.00


Check Cart for Availability


Select a Size

Change View
100 μL
$482.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$482.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

41 kDa

species reactivity

horse, pig, bovine, dog, rat, mouse, human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HEXIM1(10614)

General description

HEXIM1 is induced by hexamethylene bis-acetamide. It co-ordinates with 7SK snRNAand subsequently inhibits P-TEFb (CDK9/Cyclin T) and RNA polymerase II.
Rabbit Anti-HEXIM1 antibody recognizes bovine, human, mouse, rat, zebrafish, and canine HEXIM1.

Immunogen

Synthetic peptide directed towards the C terminal region of human HEXIM1

Application

Rabbit Anti-HEXIM1 antibody can be used for western blot applications at a concentration of 0.6 μg/ml.

Biochem/physiol Actions

HEXIM1 expression is induced by hexamethylene-bis-acetamide in vascular smooth muscle cells. The function of this protein is not yet known.

Sequence

Synthetic peptide located within the following region: LESKRLGGDDARVRELELELDRLRAENLQLLTENELHRQQERAPLSKFGD

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jasper H N Yik et al.
Molecular cell, 12(4), 971-982 (2003-10-29)
The positive transcriptional elongation factor b (P-TEFb), consisting of CDK9 and cyclin T, stimulates transcription by phosphorylating RNA polymerase II. It becomes inactivated when associated with the abundant 7SK snRNA. Here, we show that the 7SK binding alone was not

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service