Skip to Content
Merck
All Photos(1)

Key Documents

AV33727

Sigma-Aldrich

Anti-NFE2L3 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-NRF3, Anti-Nuclear factor (erythroid-derived 2)-like 3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$716.00

$716.00


Check Cart for Availability


Select a Size

Change View
100 μL
$716.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$716.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

76 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NFE2L3(9603)

General description

Nuclear factor erythroid 2-related factor 3, NFE2L3, is a basic-region leucine zipper (bZIP) transcription factor. It is a glycoprotein expressed in the endoplasmic reticulum and the nuclear membrane.

Immunogen

Synthetic peptide directed towards the middle region of human NFE2L3

Biochem/physiol Actions

NFE2L3 activates erythroid-specific, globin gene expression.

Sequence

Synthetic peptide located within the following region: NPEQTLPGTNLTGFLSPVDNHMRNLTSQDLLYDLDINIFDEINLMSLATE

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Kobayashi et al.
The Journal of biological chemistry, 274(10), 6443-6452 (1999-02-26)
The NF-E2-binding sites or Maf recognition elements (MARE) are essential cis-acting elements in the regulatory regions of erythroid-specific genes recognized by the erythroid transcription factor NF-E2, composed of p45 and MafK. Recently, two p45-related factors Nrf1 and Nrf2 were isolated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service