Skip to Content
Merck
All Photos(2)

Key Documents

AV33542

Sigma-Aldrich

Anti-ANKRD11 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Ankyrin repeat domain 11

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$547.00

$547.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1288


Select a Size

Change View
100 μL
$547.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$547.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1288

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

298 kDa

species reactivity

bovine, horse, rabbit, dog, rat, mouse, human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ANKRD11(29123)

General description

ANKRD11 is a member of a family of ankyrin repeat-containing cofactors that interacts with p160 nuclear receptor coactivators and inhibits ligand-dependent transcriptional activation by recruiting histone deacetylases to the complex. Recently, ANKRD11 was shown to bind to the tumor suppressor gene p53 and enhance its activity.

Immunogen

Synthetic peptide directed towards the N terminal region of human ANKRD11

Biochem/physiol Actions

ANKRD11 is a member of a novel family of ankyrin repeats containing cofactors (ANCOs) that interact with p160 coactivators to inhibit ligand-dependent transactivation. ANKRD11 encodes a large nuclear protein with five ankyrin repeats, and parts of its sequences have been reported as nasopharyngeal carcinoma susceptibility protein and medulloblastoma antigen. This gene also colocalizes and interacts with histone deacetylases.

Sequence

Synthetic peptide located within the following region: KRKLPFTAGANGEQKDSDTEKQGPERKRIKKEPVTRKAGLLFGMGLSGIR

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aihua Zhang et al.
The Journal of biological chemistry, 279(32), 33799-33805 (2004-06-09)
Members of the p160 nuclear receptor coactivators interact with liganded nuclear receptors to enhance transcription of target genes. Here we identify a novel family of ankyrin repeats containing cofactors (ANCOs) that interact with the p160 coactivators. ANCO-1 binds to the
Paul M Neilsen et al.
Journal of cell science, 121(Pt 21), 3541-3552 (2008-10-09)
The ability of p53 to act as a transcription factor is critical for its function as a tumor suppressor. Ankyrin repeat domain 11, ANKRD11 (also known as ANR11 or ANCO1), was found to be a novel p53-interacting protein that enhanced

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service