Skip to Content
Merck
All Photos(1)

Key Documents

AV32800

Sigma-Aldrich

Anti-PCK1 (AB1) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-MGC22652, Anti-PEPCK-C, Anti-PEPCK1, Anti-PEPCKC, Anti-Phosphoenolpyruvate carboxykinase 1 (soluble)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$490.00

$490.00


Estimated to ship on01 June 2025



Select a Size

Change View
100 μL
$490.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$490.00


Estimated to ship on01 June 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

69 kDa

species reactivity

human, bovine, rat

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PCK1(5105)

General description

PCK1 is a cytosolic enzyme that catalyzes the synthesis of phosphoenolpyruvate from oxaloacetate. Studies in mice have reported that Pck1 may have implications in diabetes and obesity-related disorders.
Rabbit Anti-PCK1 (AB1) antibody recognizes bovine, pig, human, mouse, and rat PCK1.

Immunogen

Synthetic peptide directed towards the N terminal region of human PCK1

Application

Rabbit Anti-PCK1 (AB1) antibody can be used for western blot applications at a concentration of 1μg/ml.

Biochem/physiol Actions

PCK1 is a main control point for the regulation of gluconeogenesis. PCK1, along with GTP, catalyzes the formation of phosphoenolpyruvate from oxaloacetate, with the release of carbon dioxide and GDP. The expression of PCK1 can be regulated by insulin, glucocorticoids, glucagon, cAMP, and diet. Defects in this gene are a cause of cytosolic phosphoenolpyruvate carboxykinase deficiency. This gene is a main control point for the regulation of gluconeogenesis. The cytosolic enzyme encoded by this gene, along with GTP, catalyzes the formation of phosphoenolpyruvate from oxaloacetate, with the release of carbon dioxide and GDP. The expression of this gene can be regulated by insulin, glucocorticoids, glucagon, cAMP, and diet. Defects in this gene are a cause of cytosolic phosphoenolpyruvate carboxykinase deficiency. A mitochondrial isozyme of the encoded protein also has been characterized. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications.

Sequence

Synthetic peptide located within the following region: PPQLQNGLNLSAKVVQGSLDSLPQAVREFLENNAELCQPDHIHICDGSEE

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elmus G Beale et al.
Cell biochemistry and biophysics, 48(2-3), 89-95 (2007-08-22)
The PCK1 gene (Pck1 in rodents) encodes the cytosolic isozyme of phosphoenolpyruvate carboxykinase (PEPCK-C), which is well-known for its function as a gluconeogenic enzyme in the liver and kidney. Mouse studies involving whole body and tissue-specific Pck1 knockouts as well

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service