Skip to Content
Merck
All Photos(1)

Key Documents

AV32759

Sigma-Aldrich

Anti-HMG20A (AB2) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-FLJ10739, Anti-HMGX1, Anti-High-mobility group 20A

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$537.00

$537.00


Estimated to ship on05 May 2025



Select a Size

Change View
100 μL
$537.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$537.00


Estimated to ship on05 May 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

40 kDa

species reactivity

mouse, rat, dog, guinea pig, horse, human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... HMG20A(10363)

General description

HMG20A is a homeobox gene that is expressed ubiquitously[1]. Studies in CHO-K1 cells have reported that it interacts with the poxvirus protein, CP77, and modulates its dissociation from the genome of vaccinia virus[2].
Rabbit Anti-HMG20A (AB2) antibody recognizes bovine, human, mouse, rat, and canine HMG20A.

Immunogen

Synthetic peptide directed towards the middle region of human HMG20A

Application

Rabbit Anti-HMG20A (AB2) antibody can be used for western blot applications at 1.25 μg/ml.

Biochem/physiol Actions

HMG20A plays a role in neuronal differentiation as chromatin-associated protein. HMG20A acts as inhibitor of HMG20B. HMG20A overcomes the repressive effects of the neuronal silencer REST and induces the activation of neuronal-specific genes. HMG20A involved in the recruitment of the histone methyltransferase MLL and consequent increased methylation of histone H3 lysine 4

Sequence

Synthetic peptide located within the following region: RKTQDRQKGKSHRQDAARQATHDHEKETEVKERSVFDIPIFTEEFLNHSK

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jye-Chian Hsiao et al.
Journal of virology, 80(15), 7714-7728 (2006-07-15)
Vaccinia virus does not grow in Chinese hamster ovary (CHO-K1) cells in the absence of a viral host range factor, cowpox protein CP77. In this study, CP77 was fused to the C terminus of green fluorescence protein (GFP-CP77) and a
L Sumoy et al.
Cytogenetics and cell genetics, 88(1-2), 62-67 (2000-04-25)
The HMG box encodes a conserved DNA binding domain found in many proteins and is involved in the regulation of transcription and chromatin conformation. We describe HMG20A and HMG20B, two novel human HMG box-containing genes, discovered within the EURO-IMAGE Consortium

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service