Skip to Content
Merck
All Photos(3)

Key Documents

AV32714

Sigma-Aldrich

Anti-NFIA antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Nuclear factor I/A

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$537.00

$537.00


Estimated to ship on21 April 2025



Select a Size

Change View
100 μL
$537.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$537.00


Estimated to ship on21 April 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

55 kDa

species reactivity

rat, horse, dog, bovine, human, rabbit, mouse, guinea pig

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NFIA(4774)

General description

NFIA is a transcription factor that regulates gliogenesis during spinal cord development. Haploinsufficeincy of NFIA has been linked to urinary tract defects and CNS malformation syndrome.
Rabbit Anti-NFIA recognizes chicken, canine, human, mouse, rat, zebrafish, and bovine NFIA.

Immunogen

Synthetic peptide directed towards the middle region of human NFIA

Application

Rabbit Anti-NFIA can be used for western blot applications at a concentration of 2μg/ml.

Biochem/physiol Actions

Nuclear factor I (NFI) proteins constitute a family of dimeric DNA-binding proteins with similar, and possibly identical, DNA-binding specificity. They function as cellular transcription factors and as replication factors for adenovirus DNA replication. Diversity in this protein family is generated by multiple genes, differential splicing, and heterodimerization.[supplied by OMIM].

Sequence

Synthetic peptide located within the following region: QHHRPVITGPRASPHATPSTLHFPTSPIIQQPGPYFSHPAIRYHPQETLK

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Benjamin Deneen et al.
Neuron, 52(6), 953-968 (2006-12-21)
The mechanisms controlling the transition from neurogenesis to gliogenesis in the vertebrate CNS are incompletely understood. We identified a family of transcription factors, called NFI genes, which are induced throughout the spinal cord ventricular zone (VZ) concomitantly with the induction
Weining Lu et al.
PLoS genetics, 3(5), e80-e80 (2007-05-29)
Complex central nervous system (CNS) malformations frequently coexist with other developmental abnormalities, but whether the associated defects share a common genetic basis is often unclear. We describe five individuals who share phenotypically related CNS malformations and in some cases urinary

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service