Skip to Content
Merck
All Photos(3)

Key Documents

AV32303

Sigma-Aldrich

Anti-FOXL1 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Forkhead box L1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$482.00

$482.00


Estimated to ship on05 April 2025



Select a Size

Change View
100 μL
$482.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$482.00


Estimated to ship on05 April 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

36 kDa

species reactivity

bovine, horse, human, pig, dog

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FOXL1(2300)

General description

The Forkhead Box L1 (FoxL1) is a transcription factor that modulates epithelial growth and gastrointestinal development. This transcription factor has been implicated in pancreatic and gastrointestinal carcinogenesis and can also function as prognostic biomarker for clear cell renal cell carcinoma. Studies in mce have also revealed that FoxL1 can function as a biological marker of the hepatic progenitor cells.
Rabbit Anti-FOXL1 antibody recognizes canine, human, and mouse FOXL1.

Immunogen

Synthetic peptide directed towards the N terminal region of human FOXL1

Application

Rabbit Anti-FOXL1 antibody can be used for western blot (2.5μg/ml) and IHC (4-8μg/ml) assays.

Biochem/physiol Actions

FOXL1 is a member of the forkhead family. The forkhead domain is a monomeric DNA binding motif that defines a rapidly growing family of eukaryotic transcriptional regulators. Genetic and biochemical data suggest a central role in embryonic development for forkhead proteins.

Sequence

Synthetic peptide located within the following region: HLFDPRLPALAASPMLYLYGPERPGLPLAFAPAAALAASGRAETPQKPPY

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Feng-Qiang Yang et al.
International journal of clinical and experimental pathology, 7(1), 110-122 (2014-01-16)
The Forkhead Box L1 (Foxl1) transcription factor regulates epithelial proliferation and development of gastrointestinal tract, and has been implicated in gastrointestinal and pancreatic tumorigenesis. However, the role of Foxl1 in renal cancer development and progression remains to be elucidated. The
Sara D Sackett et al.
Hepatology (Baltimore, Md.), 49(3), 920-929 (2008-12-24)
The liver contains a population of small bipotential facultative progenitor cells that reconstitute liver function when mature hepatocytes or cholangiocytes are unable to proliferate. Mesenchymal markers, including members of the forkhead transcription factor gene family, have been detected in hepatic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service