Skip to Content
Merck
All Photos(3)

Key Documents

AV32296

Sigma-Aldrich

Anti-FOXF1 (AB2) antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Forkhead box F1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$416.00

$416.00


Check Cart for Availability


Select a Size

Change View
100 μL
$416.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$416.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

38 kDa

species reactivity

horse, human, rabbit

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FOXF1(2294)

General description

FOXF1 is a transcription factor that is induced upon p53-dependent DNA damage. Studies have reported that this transcription factor may act as a p53 target. FOXF1 may also regulate the invasion and migration of cancer cells.
Rabbit Anti-FOXF1 recognizes canine, human, mouse, and chicken FOXF1.

Immunogen

Synthetic peptide directed towards the C terminal region of human FOXF1

Application

Rabbit Anti-FOXF1 (AB2) antibody can be used for western blotting assays at a concentration of 2.5μg/ml. It can also be used for IHC at 4-8μg/ml, using paraffin-embedded tissues.

Biochem/physiol Actions

This gene belongs to the forkhead family of transcription factors which is characterized by a distinct forkhead domain. The specific function of this gene has not yet been determined; however, it may play a role in the regulation of pulmonary genes as well as embryonic development.

Sequence

Synthetic peptide located within the following region: PCNPAANPLSGSLSTHSLEQPYLHQNSHNAPAELQGIPRYHSQSPSMCDR

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Tamura et al.
Oncogene, 33(40), 4837-4846 (2013-11-05)
p53 is an established tumor suppressor that can activate the transcription of multiple target genes. Recent evidence suggests that p53 may contribute to the regulation of cell invasion and migration. In this study, we show that the forkhead box transcription

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service