Skip to Content
Merck
All Photos(2)

Key Documents

AV32195

Sigma-Aldrich

Anti-DBP (AB2) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-D site of albumin promoter (albumin D-box) binding protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

34 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DBP(1628)

General description

Albumin promoter (albumin D-box) D-site binding protein (DBP), a PAR basic region leucine zipper (bZIP) protein, is a transcription factor involved in the diurnal regulation of gene transcription in the brain, liver and kidney. It regulates the expression of proteins such as cholesterol 7 α-hydroxylase P450 gene (CYP7) and kisspeptins (Kiss1).
DBP is known to control the growth of liver cells.
Rabbit polyclonal anti-DBP reacts with bovine, canine, human, mouse, and rat D-site of albumin promoter (albumin D-box) binding proteins.

Immunogen

Synthetic peptide directed towards the N terminal region of human DBP

Application

Rabbit Anti-DBP can be used for western blot applications at a concentration of 0.5-5.0μg/ml. It can also be used for immunohistochemistry at 4-8μg/ml, using paraffin-embedded tissues.
Rabbit polyclonal anti-DBP is used to tag D site of albumin promoter (albumin D-box) binding protein for detection and quantitation by immunocytochemical and immunohistochemical (IHC) techniques. It is used as a probe to determine the presence and roles of D site of albumin promoter (albumin D-box) binding protein in diurnal regulation of gene expression.

Biochem/physiol Actions

D-site-binding protein (DBP) is a member of the PAR bZIP (proline and acidic amino acid-rich basic leucine zipper) transcription factor family.

Sequence

Synthetic peptide located within the following region: PKEPASCLLKEKERKAALPAATTPGPGLETAGPADAPAGAVVGGGSPRGR

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C R Mueller et al.
Cell, 61(2), 279-291 (1990-04-20)
The full-length cDNA for a transcriptional activator, DBP, that binds to the D site of the albumin promoter has been cloned. DBP belongs to a family of related transcription factors including Fos, Jun, CREB, and C/EBP, which share a conserved

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service