Skip to Content
Merck
All Photos(2)

Key Documents

AV31990

Sigma-Aldrich

Anti-INSM1 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Insulinoma-associated 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$554.00

$554.00


Check Cart for Availability


Select a Size

Change View
100 μL
$554.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$554.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

53 kDa

species reactivity

pig, bovine, human, mouse, dog

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... INSM1(3642)

General description

Insulinoma-associated 1 (INSM1/IA-1) is a zinc-finger transcription factor restrictedly expressed in pancreatic β-cells during early pancreas development. INSM1/IA-1 regulates NeruoD1/β2 and insulin gene expression during β-cell maturation. INSM1 transcription factor, an important player in early embryonic neurogenesis, contributes to endocrine and neuroendocrine cell differentiation.
Rabbit polyclonal anti-INSM1 antibody reacts with canine, bovine, human, mouse, rat, and zebrafish Insulinoma-associated 1 transcription factors.

Immunogen

Synthetic peptide directed towards the middle region of human INSM1

Application

Rabbit Anti-INSM1 antibody can be used for western blot assays at a concentration of 1.25μg/ml. The antibody can also be used for IHC at 4-8μg/ml using paraffin-embedded tissues.
Rabbit polyclonal anti-INSM1 antibody is used to tag Insulinoma-associated 1 for detection and quantitation by immunocytochemical and immunohistochemical (IHC) techniques. It is used as a probe to determine the presence and roles of Insulinoma-associated 1 in endocrine and neuroendocrine cell differentiation.

Biochem/physiol Actions

Insulinoma-associated 1 (INSM1) gene is intronless and encodes a protein containing both a zinc finger DNA-binding domain and a putative prohormone domain. This gene is a sensitive marker for neuroendocrine differentiation of human lung tumors.

Sequence

Synthetic peptide located within the following region: PAAALRPPGKRPPPPTAAEPPAKAVKAPGAKKPKAIRKLHFEDEVTTSPV

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael S Lan et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 23(7), 2024-2033 (2009-02-28)
Zinc-finger transcription factors are DNA-binding proteins that are implicated in many diverse biological functions. INSM1 (formerly IA-1) contains five zinc-finger motifs and functions as a transcription factor. INSM1 protein structure is highly conserved in homologues of different species. It is
Wei-Dong Liu et al.
The Biochemical journal, 397(1), 169-177 (2006-03-30)
INSM1/IA-1 (insulinoma-associated 1) is a developmentally regulated zinc-finger transcription factor, exclusively expressed in the foetal pancreas and nervous system, and in tumours of neuroendocrine origin. We have identified an INSM1 binding site in the neuroD/beta2 promoter and demonstrated transcriptional repressor

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service