Skip to Content
Merck
All Photos(1)

Key Documents

AV31651

Sigma-Aldrich

Anti-ERF antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Ets2 repressor factor, Anti-PE-2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$537.00

$537.00


Estimated to ship on24 May 2025



Select a Size

Change View
100 μL
$537.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$537.00


Estimated to ship on24 May 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

59 kDa

species reactivity

horse, rat, dog, human, bovine, mouse, pig

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ERF(2077)

General description

ERF is a transcription factor that belongs to the ETS family of proteins and regulates cell growth. Reduced ERF dosage has been linked to complex craniosynostosis in humans.
Rabbit Anti-ERF antibody recognizes human, mouse, rat, and zebrafish ERF.

Immunogen

Synthetic peptide directed towards the C terminal region of human ERF

Application

Rabbit Anti-ERF antibody can be used for western blot applications at a concentration of 1μg/ml.

Biochem/physiol Actions

Members of the ETS family of transcription factors, such¡¡as ERF, regulate cell proliferation and differentiation. They share¡¡a highly conserved DNA-binding domain, the ETS domain, that¡¡recognizes the sequence GGAA/T.Members of the ETS family of transcription factors, such as ERF, regulate cell proliferation and differentiation. They share a highly conserved DNA-binding domain, the ETS domain, that recognizes the sequence GGAA/T (de Castro et al., 1997 [PubMed 9192842]). For further information on ETS transcription factors, see ETS1 (MIM 164720).[supplied by OMIM]. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications. PRIMARYREFSEQ_SPAN PRIMARY_IDENTIFIER PRIMARY_SPAN COMP 1-56 DC299706.1 1-56 57-2698 BC022231.1 1-2642 2699-2705 BM905776.1 207-213

Sequence

Synthetic peptide located within the following region: GGLAEGAGALAPPPPPPQIKVEPISEGESEEVEVTDISDEDEEDGEVFKT

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Akama et al.
Biochemical and biophysical research communications, 168(2), 857-862 (1990-04-30)
To clarify the interactions between mononuclear cells and polymorphonuclear leukocytes, and to identify the cytokine(s) that mediate the interaction, the effects of a culture supernatant of LPS-stimulated mononuclear cells on production of arachidonic acid metabolites of polymorphonuclear cells were studied.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service