Skip to Content
Merck
All Photos(1)

Documents

AV13110

Sigma-Aldrich

Anti-TRH antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-MGC125964, Anti-MGC125965, Anti-Thyrotropin-releasing hormone

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

26 kDa

species reactivity

bovine, human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TRH(7200)

General description

Rabbit polyclonal anti-TRH antibody reacts with zebrafish, human, canine, chicken, and bovine thyrotrophin-releasing hormones.
Thyrotropin-releasing hormone/thyrotropin-releasing factor (TRH, TRF) is a tripeptidal hormone that stimulates the synthesis and release of TSH (thyroid-stimulating hormone) and prolactin from the anterior pituitary. TRH is a highly specific regulator of the thyrotropin-stimulating hormone (TSHβ) gene expression in the pituitary via a nerve growth factor IB (NGFIB, NR4A1, Nur77)/PKC/ERK1/2 pathway.

Immunogen

Synthetic peptide directed towards the C terminal region of human TRH

Application

Rabbit polyclonal anti-TRH antibody is used to tag thyrotrophin-releasing hormone for detection and quantitation by immunocytochemical and immunohistochemical (IHC) techniques. It is used as a probe to determine the presence and roles of thyrotrophin-releasing hormone in the regulation of the synthesis and release of TSH (thyroid-stimulating hormone) and prolactin from the anterior pituitary. Anti-TRH antibody produced in rabbit is suitable for western blotting at a concentration of 0.5 μg/ml.

Sequence

Synthetic peptide located within the following region: RALGGPCGPQGAYGQAGLLLGLLDDLSRSQGAEEKRQHPGRRAAWVREPL

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service