Skip to Content
Merck
All Photos(1)

Key Documents

AV100932

Sigma-Aldrich

Anti-HOXA10 (AB2) antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Homeobox A10

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$487.00

$487.00


Check Cart for Availability


Select a Size

Change View
100 μL
$487.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$487.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

41 kDa

species reactivity

dog, bovine, pig, rabbit, human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HOXA10(3206)

General description

Homeobox A10 (HOXA10) is a homeodomain transcription factor involved in definitive hematopoiesis and implicated in the pathogenesis of acute myeloid leukemia (AML). HOXA10 facilitates myeloid progenitor expansion/proliferation while impeding myeloid differentiation. Sustained HoxA10expression during differentiation has been described in poor prognosis human acute myeloid leukemia (AML).
Rabbit polyclonal anti-HOXA10 antibody reacts with rabbit, pig, canine, mouse, bovine, human, and rat homeobox A10 transcription factors.

Immunogen

Synthetic peptide directed towards the N terminal region of human HOXA10

Application

Rabbit polyclonal anti-HOXA10 antibody is used to tag homeobox A10 for detection and quantitation by immunocytochemical and immunohistochemical (IHC) techniques. It is used as a probe to determine the presence and roles of homeobox A10 in hematopoiesis involving the expansion of myeloid progenitor populations and the development of acute myeloid leukemia (AML). Anti-HOXA10 (AB2) antibody produced in rabbit is suitable for western blotting at a concentration of 0.5 μg/ml.

Biochem/physiol Actions

HOXA10 is an important regulator of embryogenesis and lineage determination of hematopoietic progenitor cells. In primates, HOXA10 maintains uterine homeosis to regulate receptivity and implantation by synchronizing the maternal and embryonic signaling on the endometrial cells.

Sequence

Synthetic peptide located within the following region: SLGNSKGENAANWLTAKSGRKKRCPYTKHQTLELEKEFLFNMYLTRERRL

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fei Li et al.
Molecular medicine reports, 11(1), 509-514 (2014-10-18)
The present study aimed to investigate whether gonadotropin-releasing hormone analogues (GnRH-as), including GnRH agonists and antagonists, affect endometrial homeobox (Hox) a10 DNA methylation during the implantation window in mice. GnRH analogue mouse models were used and were treated with either
G B Godbole et al.
Reproduction (Cambridge, England), 134(3), 513-523 (2007-08-22)
Homeobox A10 (HOXA10), a member of abdominal B subclass of homeobox genes, is responsible for uterine homeosis during development. Intriguingly, in the adult murine uterus, HOXA10 has been demonstrated to play important roles in receptivity, embryo implantation, and decidualization. However
Tom Taghon et al.
Blood, 99(4), 1197-1204 (2002-02-07)
Homeobox genes are well known for their crucial role during embryogenesis but have also been found to be critically involved in normal and leukemic hematopoiesis. Because most previous studies focused on the role of aberrant HOX gene expression in leukemogenesis

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service