Skip to Content
Merck
All Photos(1)

Key Documents

AV100908

Sigma-Aldrich

Anti-CEBPA antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-CCAAT/enhancer binding protein (C/EBP), α

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$632.00

$632.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1367


Select a Size

Change View
100 μL
$632.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$632.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1367

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

38 kDa

species reactivity

pig, rat, mouse, bovine, human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CEBPA(1050)

Immunogen

Synthetic peptide directed towards the N terminal region of human CEBPA

Application

Anti-CEBPA antibody produced in rabbit is suitable for western blotting at a concentration of 5 μg/ml.

Biochem/physiol Actions

CCAAT/enhancer binding proteins are involved in the regulation of genes involved in the differentiation of squamous epithelial cells. They have been reported to exhibit altered expression in skin neoplasms. CEB proteins also mediate the functional interaction of IL-6 and TNF-α in mouse embryonic fibroblasts.

Sequence

Synthetic peptide located within the following region: GGICEHETSIDISAYIDPAAFNDEFLADLFQHSRQQEKAKAAVGPTGGGG

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matthew J Ruddy et al.
The Journal of biological chemistry, 279(4), 2559-2567 (2003-11-06)
Interleukin (IL)-17 is a recently described cytokine involved in the amplification of inflammatory responses and pathologies. A hallmark feature of IL-17 is its ability to induce expression of other cytokines and chemokines. In addition, IL-17 potently synergizes with tumor necrosis
H S Oh et al.
The Journal of investigative dermatology, 110(6), 939-945 (1998-06-10)
The epidermis is a stratified squamous epithelium composed primarily of keratinocytes that undergo sequential changes in gene expression during differentiation. CCAAT/enhancer binding proteins (C/EBP) are members of the bZIP family of DNA binding proteins/transcription factors. Northern analysis demonstrated that C/EBPalpha

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service