Skip to Content
Merck
All Photos(1)

Key Documents

AV100722

Sigma-Aldrich

Anti-DMRT1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-DMT1, Anti-Doublesex and mab-3 related transcription factor 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$558.00

$558.00


Check Cart for Availability


Select a Size

Change View
100 μL
$558.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$558.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

39 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DMRT1(1761)

Immunogen

Synthetic peptide directed towards the N terminal region of human DMRT1

Application

Anti-DMRT1 antibody produced in rabbit is suitable for western blotting at a concentration of 1 μg/ml.

Biochem/physiol Actions

DMRT1 protein is involved in sex determination, pluripotency and meiosis. Depletion of both DMRT1 and p53 in spermatogonial stem cells induces pluripotency by upregulation of Sox2. Deficiency of DMRT1 results in apoptosis of germline stem cells.

Sequence

Synthetic peptide located within the following region: PNDEAFSKPSTPSEAPHAPGVPPQGRAGGFGKASGALVGAASGSSAGGSS

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seiji Takashima et al.
Genes & development, 27(18), 1949-1958 (2013-09-14)
Spermatogonial stem cells (SSCs) present the potential to acquire pluripotency under specific culture conditions. However, the frequency of pluripotent cell derivation is low, and the mechanism of SSC reprogramming remains unknown. In this study, we report that induction of global
Ming-Hui Li et al.
Endocrinology, 154(12), 4814-4825 (2013-10-10)
Transcription activator-like effector nucleases (TALENs) are a powerful approach for targeted genome editing and have been proved to be effective in several organisms. In this study, we reported that TALENs can induce somatic mutations in Nile tilapia, an important species

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service