Skip to Content
Merck
All Photos(1)

Key Documents

AV03006

Sigma-Aldrich

Anti-CDK2 antibody produced in rabbit

IgG fraction of antiserum

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$643.00

$643.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1589


Select a Size

Change View
100 μL
$643.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$643.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1589

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

34 kDa

species reactivity

mouse, rabbit, sheep, human, bovine, goat, rat

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CDK2(1017)

Immunogen

Synthetic peptide directed towards the C terminal region of human CDK2

Application

Anti-CDK2 antibody produced in rabbit is suitable for western blotting at a concentration of 1 μg/ml. For immunohistochemistry of paraffin-embedded tissue sections, a concentration of 4-8 μg/ml is suitable.

Biochem/physiol Actions

Cyclin-dependent kinase 2 has a unique role in suppressing cell senescence and apoptosis induced by Myc. It is redundant for cell cycle progression but is activated following Myc overexpression to prevent Myc-induced senescence-like arrest. CDK2 functions in association with cyclin E and regulates meiosis at prophase I.

Sequence

Synthetic peptide located within the following region: SKVVPPLDEDGRSLLSQMLHYDPNKRISAKAALAHPFFQDVTKPVPHLRL

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stefano Campaner et al.
Cell cycle (Georgetown, Tex.), 9(18), 3655-3661 (2010-09-08)
The aberrant activation of oncogenic pathways promotes tumor progression, but concomitantly elicits compensatory tumor-suppressive responses, such as apoptosis or senescence. For example, Ras induces senescence, while Myc generally triggers apoptosis. Myc is in fact viewed as an anti-senescence oncogene, as
Per Hydbring et al.
Aging, 2(4), 244-250 (2010-05-07)
Proto-oncogenes such as MYC and RAS promote normal cell growth but fuel tumor development when deregulated. However, over-activated Myc and Ras also trigger intrinsic tumor suppressor mechanisms leading to apoptosis and senescence, respectively. When expressed together MYC and RAS are
Stefano Campaner et al.
Nature cell biology, 12(1), 54-59 (2009-12-17)
Activated oncogenes induce compensatory tumour-suppressive responses, such as cellular senescence or apoptosis, but the signals determining the main outcome remain to be fully understood. Here, we uncover a role for Cdk2 (cyclin-dependent kinase 2) in suppressing Myc-induced senescence. Short-term activation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service