Skip to Content
Merck
All Photos(1)

Key Documents

AV01011

Sigma-Aldrich

Anti-H3F3A antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Phosphohistone H3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$547.00

$547.00


Estimated to ship on31 May 2025



Select a Size

Change View
100 μL
$547.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$547.00


Estimated to ship on31 May 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

15 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... H3F3A(8290)

General description

H3F3A is a H3 histone protein that has been implicated in pediatric astrocytoma and glioblastoma. Rabbit Anti-H3F3A antibody binds to human H3F3A.

Immunogen

Synthetic peptide directed towards the N terminal region of human HIST3H3

Application

Rabbit Anti-H3F3A antibody can be used for western blot (1μg/ml) assays.

Sequence

Synthetic peptide located within the following region: MARTKQTARKSTGGKAPRKQLATKVARKSAPATGGVKKPHRYRPGTVALR

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gerrit H Gielen et al.
American journal of clinical pathology, 139(3), 345-349 (2013-02-23)
Brain tumors are one of the most common childhood malignancies. Diffuse high-grade gliomas represent approximately 10% of pediatric brain tumors. Exon sequencing has identified a mutation in K27M of the histone H3.3 gene (H3F3A K27M and G34R/V) in about 20%
Dominik Sturm et al.
Cancer cell, 22(4), 425-437 (2012-10-20)
Glioblastoma (GBM) is a brain tumor that carries a dismal prognosis and displays considerable heterogeneity. We have recently identified recurrent H3F3A mutations affecting two critical amino acids (K27 and G34) of histone H3.3 in one-third of pediatric GBM. Here, we

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service