ARLR3016A
PolarSafe™ Cardboard Freezer Box
L × W × H 5 1/4 in. × 5 1/4 in. × 3 in.
All Photos(2)
About This Item
UNSPSC Code:
41103000
Recommended Products
manufacturer/tradename
Argos R3016A
L × W × H
5 1/4 in. × 5 1/4 in. × 3 in.
General description
PolarSafe cryo/freezer boxes and dividers are made of sturdy white cardboard, with a moisture-repellent coating to protect form the effects of liquid nitrogen or condensation from refrigerated equipment. Boxes are sized to fit all standard stainless-steel and polycarbonate storage systems. Small holes on the bottom allow for safe drainage of liquid nitrogen or water. For use in mechanical freezers and liquid nitrogen tanks at temperature to -196°C.
Choice of boxes and dividers to accommodate tubes from 0.5 – 50mL.
Choice of boxes and dividers to accommodate tubes from 0.5 – 50mL.
Legal Information
PolarSafe is a trademark of Argos Technologies, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)