Merck
All Photos(4)
Documents
ARLAPS5
Serological Pipettes
capacity 5 mL, sterile, polystyrene
About This Item
UNSPSC Code:
41106514
NACRES:
NB.22
material
polystyrene
sterility
sterile
feature
individually wrapped
packaging
pkg of 200 ea
manufacturer/tradename
Argos APS5
capacity
5 mL
General description
Individually wrapped plastic pipettes that are supplied without graduations or filters for vacuum or free flow pipetting applications. Can also be used with peristaltic pumps.