Skip to Content
Merck
All Photos(1)

Key Documents

AL11480

Sigma-Aldrich

DMT-locA(bz) Phosphoramidite

configured for PerkinElmer

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$246.00

$246.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$246.00

About This Item

Empirical Formula (Hill Notation):
C48H52N7O8P
CAS Number:
Molecular Weight:
885.94
MDL number:
UNSPSC Code:
41116105
NACRES:
NA.51

$246.00


Check Cart for Availability

Request a Bulk Order

biological source

non-animal source (BSE/TSE no Risk)

Quality Level

product line

Proligo Reagents

Assay

≥98.0% (31P-NMR)
≥98.0% (reversed phase HPLC)

form

powder

impurities

≤3 wt. % Residual Solvent Content
<0.4% Water Content (Karl Fischer)
<0.5% Single unspecified Impurity (reversed phase HPLC)

color

white to off-white

solubility

soluble, clear, colorless

absorption

<0.1 in acetonitrile at 0.2 M

suitability

conforms to structure for H-NMR
conforms to structure for LC-MS

compatibility

configured for PerkinElmer

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service