Skip to Content
Merck
All Photos(1)

Documents

A9506

Sigma-Aldrich

L-Aspartic acid hemimagnesium salt hydrate

≥98% (TLC)

Synonym(s):

(S)-Aminobutanedioic acid hemimagnesium salt, Magnesium L-aspartate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[HOOCCH(NH2)CH2COO]2Mg
Molecular Weight:
144.25 (anhydrous basis)
Beilstein:
5178578
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98% (TLC)

SMILES string

N[C@@H](CC(=O)O[Mg]OC(=O)C[C@H](N)C(O)=O)C(O)=O

InChI

1S/2C4H7NO4.Mg/c2*5-2(4(8)9)1-3(6)7;/h2*2H,1,5H2,(H,6,7)(H,8,9);/q;;+2/p-2/t2*2-;/m00./s1

InChI key

RXMQCXCANMAVIO-CEOVSRFSSA-L

Gene Information

human ... GRIN2A(2903)
rat ... GRIN2A(14811)

Biochem/physiol Actions

Principal neurotransmitter for fast synaptic excitation.

replaced by

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tamami Uejima et al.
Acta crystallographica. Section D, Biological crystallography, 69(Pt 3), 345-351 (2013-03-23)
Rab small GTPases regulate vesicle transport in eukaryotes by interacting with various effectors. Guanine nucleotide-exchange factor (GEF) catalyzes the transition from inactive GDP-bound Rab to active GTP-bound Rab. The existence of several GDP-bound intermediates containing the Arabidopsis thaliana Rab5 homologue
Przemyslaw Swiatkowski et al.
Biochemical and biophysical research communications, 433(3), 354-357 (2013-03-21)
In the pharynx of Caenorhabditis elegans, the accessory subunit MPS-4, homolog to human KCNE1, forms a complex with K(+) channel EXP-2 that terminates the action potential. An aspartate residue critical for KCNE1 function, asp76, is conserved in MPS-4 (asp74). Here
Ellen W Moomaw et al.
PloS one, 8(3), e57933-e57933 (2013-03-08)
Ceriporiopsis subvermispora oxalate oxidase (CsOxOx) is the first bicupin enzyme identified that catalyzes manganese-dependent oxidation of oxalate. In previous work, we have shown that the dominant contribution to catalysis comes from the monoprotonated form of oxalate binding to a form
Ann-Louise Johansson et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(22), 8912-8917 (2013-05-16)
Proton transfer across biological membranes underpins central processes in biological systems, such as energy conservation and transport of ions and molecules. In the membrane proteins involved in these processes, proton transfer takes place through specific pathways connecting the two sides
Laureano de la Vega et al.
Nucleic acids research, 41(11), 5731-5745 (2013-04-27)
Differentiation of skeletal muscle cells is accompanied by drastic changes in gene expression programs that depend on activation and repression of genes at defined time points. Here we identify the serine/threonine kinase homeodomain-interacting protein kinase 2 (HIPK2) as a corepressor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service