A9150
Lacto-N-biose
Synonym(s):
β-D-Gal-(1→3)-D-GlcNAc, 2-Acetamido-2-deoxy-3-O-(β-D-galactopyranosyl)-D-glucopyranose
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C14H25NO11
CAS Number:
Molecular Weight:
383.35
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
Recommended Products
storage temp.
−20°C
SMILES string
CC(=O)NC(C=O)C(OC1OC(CO)C(O)C(O)C1O)C(O)C(O)CO
Biochem/physiol Actions
A disaccharide involved in the expression of Lewis blood group antigens. Possibly of interest as a lectin inhibitor.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
J P Prieels et al.
The Journal of biological chemistry, 256(20), 10456-10463 (1981-10-25)
The Lewis blood group-specified N-acetylglucosaminide alpha 1 goes to 4 fucosyltransferase and an N-acetylglucosaminide alpha 1 goes to 3 fucosyltransferase have been copurified over 500,000-fold from human milk by affinity chromatography on GDP-hexanolamine agarose. The purified enzyme preparation migrates as
T B Kuhlenschmidt et al.
Biochemistry, 23(16), 3569-3575 (1984-07-31)
Chicken hepatic lectin was isolated with affinity chromatography by using neoglycoproteins of bovine serum albumin (BSA) to which n moles of glycosides has been attached by amidination (Glycn-AI-BSA) [Lee, Y. C., Stowell, C. P., & Krantz, M. J. (1976) Biochemistry
Watkins, W.M. et al.
Glycoproteins, 830-830 (1972)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service