Skip to Content
Merck
All Photos(3)

Key Documents

A9075

Sigma-Aldrich

(2S,3R)-3-Amino-2-hydroxy-4-phenylbutyric acid hydrochloride

Synonym(s):

(2S,3R)-AHPA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13NO3 · HCl
CAS Number:
Molecular Weight:
231.68
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

solid

storage temp.

−20°C

SMILES string

Cl.N[C@H](Cc1ccccc1)[C@H](O)C(O)=O

InChI

1S/C10H13NO3.ClH/c11-8(9(12)10(13)14)6-7-4-2-1-3-5-7;/h1-5,8-9,12H,6,11H2,(H,13,14);1H/t8-,9+;/m1./s1

InChI key

OPVMPYQFOLATCK-RJUBDTSPSA-N

Biochem/physiol Actions

(2S,3R)-3-Amino-2-hydroxy-4-phenylbutyric acid (AHPA) derivatives are used as inhibitors of enkephalinase(s) to augment met5-enkephalin-induced anti-nociception. The derivative (2S,3R)-3-amino-2-hydroxy-4-phenylbutanoyl-L-valine (AHPA-Val) is used as a bestatin analogue.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Abe et al.
The Journal of antibiotics, 41(12), 1862-1867 (1988-12-01)
The biological activity of the two main metabolites of ubenimex in humans, (-)-N-[(2S,3R)-3-amino-2-hydroxy-4-(4'-hydroxy)phenylbutyryl]-L-leucine (OH-ubenimex) and (2S,3R)-3-amino-2-hydroxy-4-phenylbutyric acid [2S,3R)-AHPA) was examined. OH-Ubenimex was almost identical in inhibitory activity against mouse peritoneal resident macrophage aminopeptidases (APases) and the growth of IMC carcinoma
Y Matsuoka et al.
Japanese journal of pharmacology, 46(3), 205-210 (1988-03-01)
It has been accepted that the periaqueductal gray matter of the mid brain (PAG) and the reticular formation of the medulla oblongata in the brain stem have antinociceptive roles in the pain control pathways of mammals, and met5-enkephalin may act
M Hachisu et al.
Life sciences, 30(20), 1739-1746 (1982-05-17)
The effects of nineteen AHPA* derivatives were examined on morphine analgesia by tail-flick test in rats and on enkephalinase inhibition which was based on the formation of tyrosyl-glycyl-glycine from met-enkephalin. The correlation between the enhancement of morphine analgesia in vivo
M C Chung et al.
Bioscience, biotechnology, and biochemistry, 60(5), 898-900 (1996-05-01)
A bestatin analogue, (2S,3R)-3-amino-2-hydroxy-4-phenylbutanoyl-L-valine (AHPA-Val), from the culture filtrate of Streptomyces neyagawaensis SL-387 was obtained in a chemically defined medium containing DL-3-amino-3-phenylpropionic acid. AHPA-Val was 6 times (IC50 = 1.2 micrograms/ml) as strong as bestatin (IC50 = 7.0 micrograms/ml) against

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service