Skip to Content
Merck
All Photos(1)

Key Documents

A8676

Sigma-Aldrich

Alprenolol hydrochloride

≥98% (TLC), powder

Synonym(s):

1-(o-Allylphenoxy)-3-(isopropylamino)-2-propanol hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H23NO2 · HCl
CAS Number:
Molecular Weight:
285.81
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98% (TLC)

form

powder

color

off-white

solubility

H2O: 50 mg/mL

SMILES string

Cl[H].CC(C)NCC(O)COc1ccccc1CC=C

InChI

1S/C15H23NO2.ClH/c1-4-7-13-8-5-6-9-15(13)18-11-14(17)10-16-12(2)3;/h4-6,8-9,12,14,16-17H,1,7,10-11H2,2-3H3;1H

InChI key

RRCPAXJDDNWJBI-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Alprenolol is a nonselective β-blocker and a serotonin 5HT1A receptor antagonist.

Biochem/physiol Actions

β1- and β2-adrenoceptor antagonist.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Zhang et al.
Psychopharmacology, 132(3), 281-288 (1997-08-01)
The beta-adrenoceptor antagonist as well as serotonin 5-HT1 receptor antagonist, (-)alprenolol, was found to potentiate the disrupting effect of the noncompetitive NMDA receptor antagonist, dizocilpine, on prepulse inhibition (PPI) of the acoustic startle response (ASR) in the rat. The facilitating
Y Shirayama et al.
European journal of pharmacology, 331(2-3), 319-323 (1997-07-23)
Desipramine, imipramine, clomipramine, (-)-propranolol, (-)-alprenolol, (+/-)-pentazocine and risperidone caused a concentration-dependent inhibition of 6 nM [3H]DTG (1,3-di-o-tolylguanidine)-defined sigma (sigma) binding with Ki values of about 0.5-2.5 microM in well-washed homogenates obtained from rat cerebral cortex. The saturation studies revealed that
Marjukka Ikonen et al.
Colloids and surfaces. B, Biointerfaces, 78(2), 275-282 (2010-04-20)
In this work, isothermal titration calorimetry (ITC) combined with zeta potential measurements was used to study the binding and partitioning of three beta-blockers, alprenolol, labetalol and propranolol, and the local anaesthetic tetracaine into liposomes. The thermodynamic parameters of enthalpy, entropy
Noel Dew et al.
Colloids and surfaces. B, Biointerfaces, 70(2), 187-197 (2009-01-27)
The aim of this study was to explore if mixtures of drug containing catanionic vesicles and polymers give rise to gel formation, and if so, if drug release from these gels could be prolonged. Catanionic vesicles formed from the drug
Ron O Dror et al.
Proceedings of the National Academy of Sciences of the United States of America, 108(32), 13118-13123 (2011-07-23)
How drugs bind to their receptors--from initial association, through drug entry into the binding pocket, to adoption of the final bound conformation, or "pose"--has remained unknown, even for G-protein-coupled receptor modulators, which constitute one-third of all marketed drugs. We captured

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service