Skip to Content
Merck
All Photos(1)

Key Documents

A8353

Sigma-Aldrich

Anti-hABH3 antibody, Mouse monoclonal

clone hABH3-99, purified from hybridoma cell culture

Synonym(s):

Anti-Human AlkB Homologue 3, Monoclonal Anti-hABH3 antibody produced in mouse

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
$677.00

$677.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
200 μL
$677.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

$677.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

hABH3-99, monoclonal

form

buffered aqueous solution

mol wt

antigen ~33 kDa

species reactivity

rat, hamster, monkey, bovine, canine, human, mouse

technique(s)

immunohistochemistry: suitable
immunoprecipitation (IP): suitable
indirect ELISA: suitable
microarray: suitable
western blot: 1-2 μg/mL using total cell extract of 293T cells

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Human alkB homolog 3, alpha-ketoglutarate dependent dioxygenase (hABH3) is mainly found in the nucleoplasm and cytoplasm.[1] Anti-hABH3 antibody, Mouse monoclonal (mouse IgG1 isotype) is derived from the hABH3-99 hybridoma produced by the fusion of mouse myeloma cells (NS1 cells) and splenocytes from BALB/c mice immunized with hABH3, conjugated to keyhole limpet hemocyanin (KLH).

Specificity

Mouse Monoclonal Anti-hABH3 binds to human, monkey, bovine, dog, rat, hamster, and mouse hABH3. It does not recognize hABH1 or hABH2. The antibody epitope resides within amino acids 2-16 of human hABH3.

Application

  • Mouse Monoclonal Anti-hABH3 has been used in:
  • western blot
  • immunohistochemistry[1]
  • immunoprecipitation
  • indirect enzyme linked immunosorbent assay (ELISA)

Mouse Monoclonal Anti-hABH3 can be used for western blot (1-2μg/mL), immunohistochemistry, immunoprecipitation, indirect ELISA and microarray studies.

Biochem/physiol Actions

Human alkB homolog 3, α-ketoglutarate dependent dioxygenase (hABH3) is involved in the repair of alkylated RNA. This antibody serves as an important tool for studying the regulation of DNA/RNA repair enzymes.
hABH3 is a Fe(II)-dependent dioxygenase involved in the repair of alkylation-induced damage in DNA. hABH3 mutations have been analysed in pediatric brain tumors.
Mouse Monoclonal Anti-hABH3 binds to human, monkey, bovine, dog, rat, hamster, and mouse hABH3. It does not recognize hABH1 or hABH2. The antibody epitope resides within amino acids 2-16 of human hABH3.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Valentina Cetica et al.
Journal of neuro-oncology, 94(2), 195-201 (2009-03-18)
Alkylating agents, commonly used for brain tumor therapy, induce DNA and RNA lesions that, if not repaired, drive cells to apoptosis. Thus, cellular mechanisms that are responsible for nucleic acid repair are possibly involved in drug resistance. This work analyzes
Human and bacterial oxidative demethylases repair alkylation damage in both RNA and DNA
Aas PA, et al.
Nature, 421(6925), 859-859 (2003)
Norihisa Uemura et al.
Cancer science, 100(9), 1612-1622 (2009-06-30)
To identify the molecular background of esophageal cancer, we conducted a proteomics study using an antibody microarray consisting of 725 antibodies and surgical specimens from three cases. The microarray analysis identified 24 proteins with aberrant expression in esophageal cancer compared

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service